Product Details

SNP ID
rs115026857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:32010571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCACACGCTATAAATTTAGGTC[A/G]GCCAATAAAGTACTAACCGTATAAA
Phenotype
MIM: 602809
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF5B PubMed Links

Gene Details

Gene
KIF5B
Gene Name
kinesin family member 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004521.2 3934 UTR 3 NP_004512.1
XM_017016224.1 3934 UTR 3 XP_016871713.1

View Full Product Details