Product Details

SNP ID
rs115281814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:49456943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAACTGGAACAAGGACCACCTT[A/G]CTCCTACTGCATTATAGCTTTTGAT
Phenotype
MIM: 609413
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERCC6 PubMed Links

Gene Details

Gene
ERCC6
Gene Name
ERCC excision repair 6, chromatin remodeling factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000124.3 6530 UTR 3 NP_000115.1
NM_001277058.1 6530 Intron NP_001263987.1
NM_001277059.1 6530 Intron NP_001263988.1

View Full Product Details