Product Details

SNP ID
rs115320174
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89214185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCAATTTGTTTTTGAAGACGCC[A/G]GAAAACTATTTCATTTAACCTGAGC
Phenotype
MIM: 604551 MIM: 613497
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CH25H PubMed Links

Gene Details

Gene
CH25H
Gene Name
cholesterol 25-hydroxylase
There are no transcripts associated with this gene.

Gene
LIPA
Gene Name
lipase A, lysosomal acid type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000235.3 2147 UTR 3 NP_000226.2
NM_001127605.2 2147 UTR 3 NP_001121077.1
NM_001288979.1 2147 UTR 3 NP_001275908.1

View Full Product Details