Product Details

SNP ID
rs115475154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:27675010 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGTAATGCAGATGTTTTATGCA[C/T]AGTTTGAGTAACATAAAATAAGTTA
Phenotype
MIM: 601332
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MKX PubMed Links

Gene Details

Gene
MKX
Gene Name
mohawk homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242702.1 1503 UTR 3 NP_001229631.1
NM_173576.2 1503 UTR 3 NP_775847.2
XM_017016105.1 1503 UTR 3 XP_016871594.1
XM_017016106.1 1503 UTR 3 XP_016871595.1
XM_017016107.1 1503 Intron XP_016871596.1
XM_017016108.1 1503 UTR 3 XP_016871597.1

View Full Product Details