Product Details

SNP ID
rs115700299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:125032788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTACGGTGTGATGGGGAAAAAC[A/G]ACAGAGACATGGACACAGTCCCCAG
Phenotype
MIM: 602619
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTBP2 PubMed Links

Gene Details

Gene
CTBP2
Gene Name
C-terminal binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083914.2 Intron NP_001077383.1
NM_001290214.2 Intron NP_001277143.1
NM_001290215.2 Intron NP_001277144.1
NM_001321012.1 Intron NP_001307941.1
NM_001321013.1 Intron NP_001307942.1
NM_001321014.1 Intron NP_001307943.1
NM_001329.3 Intron NP_001320.1
NM_022802.2 Intron NP_073713.2
XM_005269561.2 Intron XP_005269618.1
XM_005269564.2 Intron XP_005269621.1
XM_005269567.2 Intron XP_005269624.1
XM_005269568.4 Intron XP_005269625.1
XM_005269569.2 Intron XP_005269626.1
XM_005269571.2 Intron XP_005269628.1
XM_005269572.3 Intron XP_005269629.1
XM_006717642.2 Intron XP_006717705.1
XM_011539349.2 Intron XP_011537651.1
XM_011539351.1 Intron XP_011537653.1
XM_011539353.1 Intron XP_011537655.1
XM_011539354.1 Intron XP_011537656.1
XM_011539355.1 Intron XP_011537657.1
XM_011539357.1 Intron XP_011537659.1
XM_011539358.1 Intron XP_011537660.1
XM_017015756.1 Intron XP_016871245.1
XM_017015757.1 Intron XP_016871246.1
Gene
MIR4296
Gene Name
microRNA 4296
There are no transcripts associated with this gene.

View Full Product Details