Product Details

SNP ID
rs115942996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97222305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGAAAACTTTAACCAAAACACT[G/T]AAGAAACCTTGTTTCTCTTGTTGAG
Phenotype
MIM: 611587
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP19 PubMed Links

Gene Details

Gene
ARHGAP19
Gene Name
Rho GTPase activating protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204300.1 5261 UTR 3 NP_001191229.1
NM_001256423.1 5261 UTR 3 NP_001243352.1
NM_032900.5 5261 UTR 3 NP_116289.4
Gene
ARHGAP19-SLIT1
Gene Name
ARHGAP19-SLIT1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details