Product Details
- SNP ID
-
rs116228045
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:93593529 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGGTTTTTTGGGGCAGTGTAGCCA[A/G]GGGTGACCACAGAAATGGCCCGAGG
- Phenotype
-
MIM: 609044
MIM: 180250
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FFAR4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs36035572] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FFAR4
- Gene Name
- free fatty acid receptor 4
There are no transcripts associated with this gene.
- Gene
- RBP4
- Gene Name
- retinol binding protein 4
View Full Product Details