Product Details

SNP ID
rs116622573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:27110491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAGATAATTTTAATGAAGTCAAC[A/G]TCAAGAATATAATCTGGTTGTAATA
Phenotype
MIM: 610855 MIM: 607472
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD26 PubMed Links

Gene Details

Gene
ANKRD26
Gene Name
ankyrin repeat domain 26
There are no transcripts associated with this gene.

Gene
YME1L1
Gene Name
YME1 like 1 ATPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253866.1 4301 UTR 3 NP_001240795.1
NM_014263.3 4301 UTR 3 NP_055078.1
NM_139312.2 4301 UTR 3 NP_647473.1
XM_011519300.2 4301 UTR 3 XP_011517602.1

View Full Product Details