Product Details

SNP ID
rs116643161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:80538554 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCACAGAGAGGTAGGCCACCAGT[C/T]CCCCAGGAGGTAGAAAAATTAGCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929574 PubMed Links

Gene Details

Gene
LOC101929574
Gene Name
uncharacterized LOC101929574
There are no transcripts associated with this gene.

Gene
LOC102723703
Gene Name
uncharacterized LOC102723703
There are no transcripts associated with this gene.

Gene
SH2D4B
Gene Name
SH2 domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145719.1 653 Intron NP_001139191.1
NM_207372.2 653 Intron NP_997255.2
XM_017016236.1 653 UTR 5 XP_016871725.1

View Full Product Details