Product Details

SNP ID
rs117039173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:127913920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCATGGGGTAAAGCTTTTTATT[C/T]TGTTGTTGGAAACACAGTATATGGT
Phenotype
MIM: 600926
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTPRE PubMed Links

Gene Details

Gene
PTPRE
Gene Name
protein tyrosine phosphatase, receptor type E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316676.1 Intron NP_001303605.1
NM_001316677.1 Intron NP_001303606.1
NM_001323354.1 Intron NP_001310283.1
NM_001323355.1 Intron NP_001310284.1
NM_001323356.1 Intron NP_001310285.1
NM_001323357.1 Intron NP_001310286.1
NM_006504.5 Intron NP_006495.1
NM_130435.4 Intron NP_569119.1
XM_005252691.2 Intron XP_005252748.1
XM_011539994.2 Intron XP_011538296.1
XM_011539995.1 Intron XP_011538297.1
XM_011539996.1 Intron XP_011538298.1
XM_011539998.1 Intron XP_011538300.1
XM_017016467.1 Intron XP_016871956.1
XM_017016468.1 Intron XP_016871957.1
XM_017016469.1 Intron XP_016871958.1

View Full Product Details