Product Details

SNP ID
rs117131154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102109525 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACAGAGCTAGGGGTAGACAAGGA[A/G]GTGGCTTGTCAGGGGACAGACATGG
Phenotype
MIM: 603451
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LDB1 PubMed Links

Gene Details

Gene
LDB1
Gene Name
LIM domain binding 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113407.2 Intron NP_001106878.1
NM_001321612.1 Intron NP_001308541.1
NM_003893.4 Intron NP_003884.1
XM_017016867.1 Intron XP_016872356.1
XM_017016868.1 Intron XP_016872357.1
XM_017016869.1 Intron XP_016872358.1

View Full Product Details