Product Details

SNP ID
rs117145794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:43649179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCAGGGACTCAGGAATACAGGG[C/T]TCTTGCAGGGCGTCGCGGAAGGAAA
Phenotype
MIM: 194539
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF32 PubMed Links

Gene Details

Gene
ZNF32
Gene Name
zinc finger protein 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005368.2 172 Intron NP_001005368.1
NM_001324164.1 172 Intron NP_001311093.1
NM_001324165.1 172 Intron NP_001311094.1
NM_001324166.1 172 Intron NP_001311095.1
NM_001324167.1 172 Intron NP_001311096.1
NM_001324168.1 172 Intron NP_001311097.1
NM_006973.2 172 Intron NP_008904.1
XM_017016616.1 172 UTR 5 XP_016872105.1
Gene
ZNF32-AS1
Gene Name
ZNF32 antisense RNA 1
There are no transcripts associated with this gene.

Gene
ZNF32-AS2
Gene Name
ZNF32 antisense RNA 2
There are no transcripts associated with this gene.

Gene
ZNF32-AS3
Gene Name
ZNF32 antisense RNA 3
There are no transcripts associated with this gene.

View Full Product Details