Product Details
- SNP ID
-
rs117193245
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:38115260 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAGGCGAGGAGCCATGGATATTA[C/G]AGGAAAAATTTCCAAGCCAGAGTCA
- Phenotype
-
MIM: 616085
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF37A
PubMed Links
Gene Details
- Gene
- ZNF37A
- Gene Name
- zinc finger protein 37A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001007094.2 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
NP_001007095.1 |
NM_001178101.1 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
NP_001171572.1 |
NM_001324245.1 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
NP_001311174.1 |
NM_001324246.1 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
NP_001311175.1 |
NM_001324247.1 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
NP_001311176.1 |
NM_001324248.1 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
NP_001311177.1 |
NM_001324249.1 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
NP_001311178.1 |
NM_001324250.1 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
NP_001311179.1 |
NM_001324251.1 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
NP_001311180.1 |
NM_001324252.1 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
NP_001311181.1 |
NM_001324253.1 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
NP_001311182.1 |
NM_001324254.1 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
NP_001311183.1 |
NM_001324256.1 |
768 |
Intron |
|
|
NP_001311185.1 |
NM_001324257.1 |
768 |
Intron |
|
|
NP_001311186.1 |
NM_001324258.1 |
768 |
Intron |
|
|
NP_001311187.1 |
NM_001324259.1 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
NP_001311188.1 |
NM_001324260.1 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
NP_001311189.1 |
NM_001324261.1 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
NP_001311190.1 |
NM_001324262.1 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
NP_001311191.1 |
NM_003421.2 |
768 |
Intron |
|
|
NP_003412.1 |
XM_005252586.3 |
768 |
Intron |
|
|
XP_005252643.1 |
XM_011519656.2 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
XP_011517958.1 |
XM_011519657.2 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
XP_011517959.1 |
XM_011519658.2 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
XP_011517960.1 |
XM_017016618.1 |
768 |
UTR 5 |
|
|
XP_016872107.1 |
XM_017016619.1 |
768 |
UTR 5 |
|
|
XP_016872108.1 |
XM_017016620.1 |
768 |
UTR 5 |
|
|
XP_016872109.1 |
XM_017016621.1 |
768 |
Intron |
|
|
XP_016872110.1 |
XM_017016622.1 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
XP_016872111.1 |
XM_017016623.1 |
768 |
Intron |
|
|
XP_016872112.1 |
XM_017016624.1 |
768 |
Missense Mutation |
CAG,GAG |
Q70E |
XP_016872113.1 |
View Full Product Details