Product Details

SNP ID
rs117574893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:53803542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATGACATATCACATATAAAATT[C/T]AAGGCAAACTTTCTTCTGTATTTTA
Phenotype
MIM: 605514
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCDH15 PubMed Links

Gene Details

Gene
PCDH15
Gene Name
protocadherin related 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142763.1 8481 Intron NP_001136235.1
NM_001142764.1 8481 Intron NP_001136236.1
NM_001142765.1 8481 Intron NP_001136237.1
NM_001142766.1 8481 Intron NP_001136238.1
NM_001142767.1 8481 Intron NP_001136239.1
NM_001142768.1 8481 Intron NP_001136240.1
NM_001142769.1 8481 Intron NP_001136241.1
NM_001142770.1 8481 Intron NP_001136242.1
NM_001142771.1 8481 UTR 3 NP_001136243.1
NM_001142772.1 8481 UTR 3 NP_001136244.1
NM_001142773.1 8481 Intron NP_001136245.1
NM_033056.3 8481 Intron NP_149045.3
XM_017016571.1 8481 Intron XP_016872060.1
XM_017016572.1 8481 UTR 3 XP_016872061.1
XM_017016573.1 8481 Intron XP_016872062.1

View Full Product Details