Product Details

SNP ID
rs118014822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:71750845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCACGGGGGAGAAGTCTCTCCAC[C/T]GTGCCCTGTGTCTGGTGCCTGGGAG
Phenotype
MIM: 615608 MIM: 605516
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf54 PubMed Links

Gene Details

Gene
C10orf54
Gene Name
chromosome 10 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022153.1 1485 UTR 3 NP_071436.1
Gene
CDH23
Gene Name
cadherin-related 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171930.1 1485 Intron NP_001165401.1
NM_001171931.1 1485 Intron NP_001165402.1
NM_001171932.1 1485 Intron NP_001165403.1
NM_001171933.1 1485 Intron NP_001165404.1
NM_001171934.1 1485 Intron NP_001165405.1
NM_001171935.1 1485 Intron NP_001165406.1
NM_001171936.1 1485 Intron NP_001165407.1
NM_022124.5 1485 Intron NP_071407.4
NM_052836.3 1485 Intron NP_443068.1
XM_006717940.3 1485 Intron XP_006718003.1
XM_006717942.3 1485 Intron XP_006718005.1
XM_011540039.2 1485 Intron XP_011538341.1
XM_011540042.2 1485 Intron XP_011538344.1
XM_011540043.2 1485 Intron XP_011538345.1
XM_011540044.2 1485 Intron XP_011538346.1
XM_011540045.2 1485 Intron XP_011538347.1
XM_011540046.2 1485 Intron XP_011538348.1
XM_011540047.2 1485 Intron XP_011538349.1
XM_011540048.2 1485 Intron XP_011538350.1
XM_011540049.2 1485 Intron XP_011538351.1
XM_011540051.2 1485 Intron XP_011538353.1
XM_011540052.2 1485 Intron XP_011538354.1
XM_017016499.1 1485 Intron XP_016871988.1
XM_017016500.1 1485 Intron XP_016871989.1
XM_017016501.1 1485 Intron XP_016871990.1
XM_017016502.1 1485 Intron XP_016871991.1
XM_017016503.1 1485 Intron XP_016871992.1
XM_017016504.1 1485 Intron XP_016871993.1
XM_017016505.1 1485 Intron XP_016871994.1
XM_017016506.1 1485 Intron XP_016871995.1
XM_017016507.1 1485 Intron XP_016871996.1
XM_017016508.1 1485 Intron XP_016871997.1

View Full Product Details