Product Details

SNP ID
rs118152880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:112286166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTGTCAACTACTCCTTCTCCTG[C/T]ACCTACCACTCCACCTACTTGGTGA
Phenotype
MIM: 602653
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TECTB PubMed Links

Gene Details

Gene
TECTB
Gene Name
tectorin beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_058222.2 436 Silent Mutation TGC,TGT C121C NP_478129.1
XM_017016604.1 436 Silent Mutation TGC,TGT C121C XP_016872093.1

View Full Product Details