Product Details

SNP ID
rs118178925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102396985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACCTCGTGCTCATGCCCACAGT[C/T]TGGTGGGCAAGCAATGCTCGGAGCT
Phenotype
MIM: 164012 MIM: 602327
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NFKB2 PubMed Links

Gene Details

Gene
NFKB2
Gene Name
nuclear factor kappa B subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077494.3 598 Silent Mutation CTG,TTG L109L NP_001070962.1
NM_001261403.2 598 Silent Mutation CTG,TTG L109L NP_001248332.1
NM_001288724.1 598 Silent Mutation CTG,TTG L109L NP_001275653.1
NM_001322934.1 598 Silent Mutation CTG,TTG L109L NP_001309863.1
NM_001322935.1 598 Silent Mutation CTG,TTG L109L NP_001309864.1
NM_002502.5 598 Silent Mutation CTG,TTG L109L NP_002493.3
XM_011539830.2 598 Intron XP_011538132.1
XM_011539831.2 598 Intron XP_011538133.1
XM_017016278.1 598 Silent Mutation CTG,TTG L290L XP_016871767.1
Gene
PSD
Gene Name
pleckstrin and Sec7 domain containing
There are no transcripts associated with this gene.

View Full Product Details