Product Details

SNP ID
rs137852662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101774771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCTGCCATGGCGTTGATGCGCT[T/C]GTTGGCCAGGACCTGCACGTGCTTC
Phenotype
MIM: 600483 MIM: 604039 MIM: 606456
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
FGF8 PubMed Links

Gene Details

Gene
FGF8
Gene Name
fibroblast growth factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206389.1 400 UTR 5 NP_001193318.1
NM_006119.4 400 Missense Mutation AAG,GAG K71E NP_006110.1
NM_033163.3 400 Missense Mutation AAG,GAG K100E NP_149353.1
NM_033164.3 400 Missense Mutation AAG,GAG K89E NP_149354.1
NM_033165.3 400 Missense Mutation AAG,GAG K60E NP_149355.1
Gene
MGEA5
Gene Name
meningioma expressed antigen 5 (hyaluronidase)
There are no transcripts associated with this gene.

Gene
NPM3
Gene Name
nucleophosmin/nucleoplasmin 3
There are no transcripts associated with this gene.

View Full Product Details