Product Details

SNP ID
rs137946307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:56358596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTCTCTCCCCATGGTGTCTCCT[A/G]TACTCTGCTCCTGGGGTCGAGTCGG
Phenotype
MIM: 609177
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZWINT PubMed Links

Gene Details

Gene
ZWINT
Gene Name
ZW10 interacting kinetochore protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005413.1 789 Missense Mutation ACA,ATA T204I NP_001005413.1
NM_007057.3 789 Missense Mutation ACA,ATA T251I NP_008988.2
NM_032997.2 789 Missense Mutation ACA,ATA T251I NP_127490.1
XM_017015605.1 789 Nonsense Mutation CAG,TAG Q239* XP_016871094.1

View Full Product Details