Product Details

SNP ID
rs138078604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:72898815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGCATGGCGGGAGATGCCATGCC[A/T]ACCTTCTGCATACCAGAAAACCACT
Phenotype
MIM: 609330
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OIT3 PubMed Links

Gene Details

Gene
OIT3
Gene Name
oncoprotein induced transcript 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152635.2 456 Silent Mutation CCA,CCT P71P NP_689848.1

View Full Product Details