Product Details

SNP ID
rs138091745
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5768297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTTCTTGATGGGGTGGCTGAGGA[C/T]GCAAATAACTCTGATCACCTGGCCC
Phenotype
MIM: 600767
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM208B PubMed Links

Gene Details

Gene
FAM208B
Gene Name
family with sequence similarity 208 member B
There are no transcripts associated with this gene.

Gene
GDI2
Gene Name
GDP dissociation inhibitor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001115156.1 893 Missense Mutation ATC,GTC I258V NP_001108628.1
NM_001494.3 893 Missense Mutation ATC,GTC I303V NP_001485.2
XM_017016071.1 893 Missense Mutation ATC,GTC I281V XP_016871560.1

View Full Product Details