Product Details

SNP ID
rs138431351
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73243491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCATCTGAGCCAGAAAATTGTCC[A/G]TTTCCTTTTGCCGATCCTTTTGTCT
Phenotype
MIM: 611206 MIM: 609204
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC9 PubMed Links

Gene Details

Gene
DNAJC9
Gene Name
DnaJ heat shock protein family (Hsp40) member C9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015190.4 834 Missense Mutation ACG,ATG T231M NP_056005.1
Gene
DNAJC9-AS1
Gene Name
DNAJC9 antisense RNA 1
There are no transcripts associated with this gene.

Gene
FAM149B1
Gene Name
family with sequence similarity 149 member B1
There are no transcripts associated with this gene.

Gene
MRPS16
Gene Name
mitochondrial ribosomal protein S16
There are no transcripts associated with this gene.

View Full Product Details