Product Details

SNP ID
rs138471857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:35640239 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGTAGACCAGCAAGAAGACCAC[C/G]GTGCACAGCGCGGGGCCGGTGGTCT
Phenotype
MIM: 606146
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FZD8 PubMed Links

Gene Details

Gene
FZD8
Gene Name
frizzled class receptor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031866.2 1196 Silent Mutation ACC,ACG T397T NP_114072.1
Gene
MIR4683
Gene Name
microRNA 4683
There are no transcripts associated with this gene.

View Full Product Details