Product Details

SNP ID
rs138484398
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133381502 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAACTGCATGATGGCCGCCCTGCC[A/G]GAGGACACTGTAGTTTTTGAGAAGC
Phenotype
MIM: 602292 MIM: 615853
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ECHS1 PubMed Links

Gene Details

Gene
ECHS1
Gene Name
enoyl-CoA hydratase, short chain 1
There are no transcripts associated with this gene.

Gene
MIR3944
Gene Name
microRNA 3944
There are no transcripts associated with this gene.

Gene
PAOX
Gene Name
polyamine oxidase (exo-N4-amino)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152911.3 791 Silent Mutation CCA,CCG P237P NP_690875.1
NM_207127.2 791 Silent Mutation CCA,CCG P237P NP_997010.1
NM_207128.2 791 Silent Mutation CCA,CCG P237P NP_997011.1

View Full Product Details