Product Details
- SNP ID
-
rs138637203
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:13439178 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAAATAAGCAAGATAATTTCAGAG[A/G]GAGAGGCAAGAGATGTGAAGGGTAA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BEND7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10590401] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BEND7
- Gene Name
- BEN domain containing 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001100912.1 |
1486 |
Intron |
|
|
NP_001094382.1 |
NM_152751.2 |
1486 |
Silent Mutation |
TCC,TCT |
S467S |
NP_689964.2 |
XM_005252402.4 |
1486 |
Silent Mutation |
TCC,TCT |
S519S |
XP_005252459.1 |
XM_011519390.2 |
1486 |
Silent Mutation |
TCC,TCT |
S480S |
XP_011517692.1 |
XM_011519391.2 |
1486 |
Silent Mutation |
TCC,TCT |
S479S |
XP_011517693.1 |
XM_011519392.2 |
1486 |
Silent Mutation |
TCC,TCT |
S518S |
XP_011517694.1 |
XM_011519393.2 |
1486 |
Silent Mutation |
TCC,TCT |
S504S |
XP_011517695.1 |
XM_011519394.2 |
1486 |
Silent Mutation |
TCC,TCT |
S480S |
XP_011517696.1 |
XM_011519395.2 |
1486 |
Silent Mutation |
TCC,TCT |
S480S |
XP_011517697.1 |
XM_011519396.2 |
1486 |
Silent Mutation |
TCC,TCT |
S480S |
XP_011517698.1 |
XM_011519397.2 |
1486 |
Silent Mutation |
TCC,TCT |
S480S |
XP_011517699.1 |
XM_011519398.2 |
1486 |
Intron |
|
|
XP_011517700.1 |
XM_011519399.2 |
1486 |
Intron |
|
|
XP_011517701.1 |
XM_011519400.2 |
1486 |
Intron |
|
|
XP_011517702.1 |
XM_011519401.2 |
1486 |
Intron |
|
|
XP_011517703.1 |
XM_011519402.2 |
1486 |
Intron |
|
|
XP_011517704.1 |
XM_011519403.2 |
1486 |
Intron |
|
|
XP_011517705.1 |
XM_011519404.2 |
1486 |
Intron |
|
|
XP_011517706.1 |
XM_011519405.2 |
1486 |
Intron |
|
|
XP_011517707.1 |
XM_011519406.2 |
1486 |
Intron |
|
|
XP_011517708.1 |
XM_011519407.1 |
1486 |
Intron |
|
|
XP_011517709.1 |
XM_011519408.2 |
1486 |
Intron |
|
|
XP_011517710.1 |
XM_011519410.2 |
1486 |
Intron |
|
|
XP_011517712.1 |
XM_011519411.2 |
1486 |
Intron |
|
|
XP_011517713.1 |
XM_011519412.1 |
1486 |
Intron |
|
|
XP_011517714.1 |
XM_011519413.1 |
1486 |
Intron |
|
|
XP_011517715.1 |
XM_011519414.1 |
1486 |
Intron |
|
|
XP_011517716.1 |
XM_017015915.1 |
1486 |
Silent Mutation |
TCC,TCT |
S480S |
XP_016871404.1 |
XM_017015916.1 |
1486 |
Intron |
|
|
XP_016871405.1 |
XM_017015917.1 |
1486 |
Intron |
|
|
XP_016871406.1 |
XM_017015918.1 |
1486 |
Intron |
|
|
XP_016871407.1 |
XM_017015919.1 |
1486 |
Intron |
|
|
XP_016871408.1 |
View Full Product Details