Product Details

SNP ID
rs138717832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:132785029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTCGTCCGAGTTGGGGTCCAGGG[A/G]CCGGTTGTATTCGTCGTCGTCCTCC
Phenotype
MIM: 600106 MIM: 605955
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INPP5A PubMed Links

Gene Details

Gene
INPP5A
Gene Name
inositol polyphosphate-5-phosphatase A
There are no transcripts associated with this gene.

Gene
NKX6-2
Gene Name
NK6 homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177400.2 800 Missense Mutation NP_796374.1
XM_017016789.1 800 Missense Mutation XP_016872278.1

View Full Product Details