Product Details

SNP ID
rs138921995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:86937085 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAACTTCACTGTCTGCAGGGCAGC[C/T]GTCCCTTCTGAAAAGCTGGCATAGA
Phenotype
MIM: 601299 MIM: 608925
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BMPR1A PubMed Links

Gene Details

Gene
BMPR1A
Gene Name
bone morphogenetic protein receptor type 1A
There are no transcripts associated with this gene.

Gene
MMRN2
Gene Name
multimerin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024756.2 2674 Silent Mutation ACA,ACG T836T NP_079032.2
XM_005270153.1 2674 Silent Mutation ACA,ACG T775T XP_005270210.1
XM_006717970.3 2674 Silent Mutation ACA,ACG T775T XP_006718033.1

View Full Product Details