Product Details

SNP ID
rs138933816
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:96981682 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAGAAATAAGGTGGGTTACCAT[C/T]TACATCCCAGTGATAAGGGCCAGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf12 PubMed Links

Gene Details

Gene
C10orf12
Gene Name
chromosome 10 open reading frame 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015652.2 399 Silent Mutation CTA,TTA L98L NP_056467.2

View Full Product Details