Product Details

SNP ID
rs138953644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89402283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATCTGTTTTTGTTTTTACAGTA[A/C]AAATGGTGATGATCATCAGGTCAAG
Phenotype
MIM: 147690
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IFIT1 PubMed Links

Gene Details

Gene
IFIT1
Gene Name
interferon induced protein with tetratricopeptide repeats 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270927.1 284 Missense Mutation AAA,ACA K3T NP_001257856.1
NM_001270928.1 284 UTR 5 NP_001257857.1
NM_001270929.1 284 UTR 5 NP_001257858.1
NM_001270930.1 284 UTR 5 NP_001257859.1
NM_001548.4 284 Missense Mutation AAA,ACA K3T NP_001539.3

View Full Product Details