Product Details

SNP ID
rs139060598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:30845065 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCACAGTTATGCTGGTGCTTCC[A/G]GTGGTGGAGGAGGTCCTCTTTCCGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF438 PubMed Links

Gene Details

Gene
ZNF438
Gene Name
zinc finger protein 438
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143766.1 2858 Missense Mutation CGG,TGG R795W NP_001137238.1
NM_001143767.1 2858 Missense Mutation CGG,TGG R795W NP_001137239.1
NM_001143768.1 2858 Missense Mutation CGG,TGG R795W NP_001137240.1
NM_001143769.1 2858 Missense Mutation CGG,TGG R746W NP_001137241.1
NM_001143770.1 2858 Missense Mutation CGG,TGG R785W NP_001137242.1
NM_001143771.1 2858 Missense Mutation CGG,TGG R785W NP_001137243.1
NM_182755.2 2858 Missense Mutation CGG,TGG R795W NP_877432.1
XM_006717393.3 2858 Missense Mutation CGG,TGG R795W XP_006717456.1
XM_006717396.3 2858 Missense Mutation CGG,TGG R795W XP_006717459.1
XM_006717398.3 2858 Missense Mutation CGG,TGG R785W XP_006717461.1
XM_006717399.3 2858 Missense Mutation CGG,TGG R785W XP_006717462.1
XM_011519371.2 2858 Missense Mutation CGG,TGG R795W XP_011517673.1
XM_011519372.2 2858 Missense Mutation CGG,TGG R795W XP_011517674.1
XM_011519373.2 2858 Missense Mutation CGG,TGG R795W XP_011517675.1
XM_011519374.2 2858 Missense Mutation CGG,TGG R795W XP_011517676.1
XM_011519376.2 2858 Missense Mutation CGG,TGG R785W XP_011517678.1
XM_011519377.2 2858 Missense Mutation CGG,TGG R785W XP_011517679.1
XM_017015863.1 2858 Missense Mutation CGG,TGG R806W XP_016871352.1
XM_017015864.1 2858 Missense Mutation CGG,TGG R795W XP_016871353.1
XM_017015865.1 2858 Missense Mutation CGG,TGG R795W XP_016871354.1
XM_017015866.1 2858 Missense Mutation CGG,TGG R795W XP_016871355.1
XM_017015867.1 2858 Missense Mutation CGG,TGG R795W XP_016871356.1
XM_017015868.1 2858 Missense Mutation CGG,TGG R795W XP_016871357.1
XM_017015869.1 2858 Missense Mutation CGG,TGG R795W XP_016871358.1
XM_017015870.1 2858 Missense Mutation CGG,TGG R795W XP_016871359.1
XM_017015871.1 2858 Missense Mutation CGG,TGG R795W XP_016871360.1
XM_017015872.1 2858 Missense Mutation CGG,TGG R795W XP_016871361.1
XM_017015873.1 2858 Missense Mutation CGG,TGG R785W XP_016871362.1
XM_017015874.1 2858 Missense Mutation CGG,TGG R785W XP_016871363.1
XM_017015875.1 2858 Missense Mutation CGG,TGG R359W XP_016871364.1
XM_017015876.1 2858 Missense Mutation CGG,TGG R359W XP_016871365.1
XM_017015877.1 2858 Missense Mutation CGG,TGG R359W XP_016871366.1
XM_017015878.1 2858 Missense Mutation CGG,TGG R359W XP_016871367.1
XM_017015879.1 2858 Missense Mutation CGG,TGG R359W XP_016871368.1
XM_017015880.1 2858 Missense Mutation CGG,TGG R359W XP_016871369.1
XM_017015881.1 2858 Missense Mutation CGG,TGG R359W XP_016871370.1
XM_017015882.1 2858 Missense Mutation CGG,TGG R359W XP_016871371.1

View Full Product Details