Product Details

SNP ID
rs139131012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:104122975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTTCTTTTTTAGAGAAAAACCA[A/G]GATTTCACTTTCAAGATGGAAAGTC
Phenotype
MIM: 616527
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP43 PubMed Links

Gene Details

Gene
CFAP43
Gene Name
cilia and flagella associated protein 43
There are no transcripts associated with this gene.

Gene
SFR1
Gene Name
SWI5 dependent homologous recombination repair protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002759.1 75 Silent Mutation CAA,CAG Q8Q NP_001002759.1
NM_145247.4 75 UTR 5 NP_660290.3
XM_005269521.3 75 Silent Mutation CAA,CAG Q70Q XP_005269578.1
XM_017015672.1 75 UTR 5 XP_016871161.1

View Full Product Details