Product Details

SNP ID
rs139138878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73134792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATCTTCCTCATCTGAATTGTTAT[C/T]GGTAGTCTGGGATACAGGTTCCTTA
Phenotype
MIM: 616464 MIM: 609233
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ECD PubMed Links

Gene Details

Gene
ECD
Gene Name
ecdysoneless cell cycle regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135752.1 2068 Missense Mutation AAT,GAT N609D NP_001129224.1
NM_001135753.1 2068 Missense Mutation AAT,GAT N533D NP_001129225.1
NM_007265.2 2068 Missense Mutation AAT,GAT N576D NP_009196.1
Gene
NUDT13
Gene Name
nudix hydrolase 13
There are no transcripts associated with this gene.

View Full Product Details