Product Details

SNP ID
rs139203737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:72274812 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAGGAGAGCCTGGCCCAGGCGC[A/G]GCTGGGCTCTCGACGCCCTGCGCGC
Phenotype
MIM: 607729
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDIT4 PubMed Links

Gene Details

Gene
DDIT4
Gene Name
DNA damage inducible transcript 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019058.2 525 Missense Mutation CAG,CGG Q108R NP_061931.1

View Full Product Details