Product Details

SNP ID
rs139248125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:72099965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTCCGTAGCAATATGGTATTTC[C/T]GTATGGTATTTCTTATGATGTTTTT
Phenotype
MIM: 614215
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASCC1 PubMed Links

Gene Details

Gene
ASCC1
Gene Name
activating signal cointegrator 1 complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198798.2 Intron NP_001185727.1
NM_001198799.2 Intron NP_001185728.1
NM_001198800.2 Intron NP_001185729.1
XM_006717873.3 Intron XP_006717936.1
XM_011539841.2 Intron XP_011538143.1
XM_011539842.2 Intron XP_011538144.1
XM_017016291.1 Intron XP_016871780.1
XM_017016292.1 Intron XP_016871781.1
XM_017016293.1 Intron XP_016871782.1
XM_017016294.1 Intron XP_016871783.1
XM_017016295.1 Intron XP_016871784.1
XM_017016296.1 Intron XP_016871785.1
XM_017016297.1 Intron XP_016871786.1
XM_017016298.1 Intron XP_016871787.1
XM_017016299.1 Intron XP_016871788.1
XM_017016300.1 Intron XP_016871789.1
XM_017016301.1 Intron XP_016871790.1
XM_017016302.1 Intron XP_016871791.1
XM_017016303.1 Intron XP_016871792.1

View Full Product Details