Product Details

SNP ID
rs139264891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:99877332 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTTGAAGGTGTAGACAGCAAAA[C/T]AGACCTGTGTGAGGGAGAGAGAGAA
Phenotype
MIM: 611282
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNMBP PubMed Links

Gene Details

Gene
DNMBP
Gene Name
dynamin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318326.1 4545 Missense Mutation TAT,TGT Y1150C NP_001305255.1
NM_001318327.1 4545 Missense Mutation TAT,TGT Y806C NP_001305256.1
NM_015221.3 4545 Missense Mutation TAT,TGT Y1518C NP_056036.1
XM_006717735.2 4545 Missense Mutation TAT,TGT Y1475C XP_006717798.1
XM_011539559.2 4545 Missense Mutation TAT,TGT Y1518C XP_011537861.1

View Full Product Details