Product Details

SNP ID
rs139387078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:68231406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTCGGCCAGGATCCGGGTCAGA[A/G]CCATGATGTAGCTCAGGGCCATCTG
Phenotype
MIM: 609875
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATOH7 PubMed Links

Gene Details

Gene
ATOH7
Gene Name
atonal bHLH transcription factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145178.3 708 Missense Mutation GCT,GTT A91V NP_660161.1

View Full Product Details