Product Details
- SNP ID
-
rs139405842
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:80142102 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACAGCTGCCGAACCCTTCAGCCCT[C/G]CGCGAGGAGACTCAGCTCAGAGCAC
- Phenotype
-
MIM: 612857
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PLAC9
PubMed Links
Gene Details
- Gene
- PLAC9
- Gene Name
- placenta specific 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001012973.1 |
346 |
Missense Mutation |
CCG,GCG |
P29A |
NP_001012991.1 |
View Full Product Details