Product Details

SNP ID
rs139440669
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102919845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACGTGATGACCCCACTCCGGGA[C/T]TGCTTCATGATCACCGGCGAAGCCA
Phenotype
MIM: 607803
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNNM2 PubMed Links

Gene Details

Gene
CNNM2
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017649.4 1553 Silent Mutation GAC,GAT D455D NP_060119.3
NM_199076.2 1553 Silent Mutation GAC,GAT D455D NP_951058.1
NM_199077.2 1553 Silent Mutation GAC,GAT D455D NP_951059.1
XM_005269933.4 1553 Silent Mutation GAC,GAT D455D XP_005269990.1
XM_006717908.2 1553 Silent Mutation GAC,GAT D455D XP_006717971.1
XM_011539911.2 1553 Silent Mutation GAC,GAT D455D XP_011538213.1
Gene
LOC107984265
Gene Name
translation initiation factor IF-2-like
There are no transcripts associated with this gene.

View Full Product Details