Product Details

SNP ID
rs139514719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:58388693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATATATCAAGATGCTTATAGGGC[A/G]GAGTGGCAGGTATATAAAGAAGAGA
Phenotype
MIM: 600438
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TFAM PubMed Links

Gene Details

Gene
TFAM
Gene Name
transcription factor A, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270782.1 720 Silent Mutation GCA,GCG A105A NP_001257711.1
NM_003201.2 720 Silent Mutation GCA,GCG A105A NP_003192.1
XM_011540120.2 720 Silent Mutation GCA,GCG A105A XP_011538422.1
XM_011540121.2 720 Silent Mutation GCA,GCG A105A XP_011538423.1

View Full Product Details