Product Details
- SNP ID
-
rs139568157
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:5499265 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAAGCTGATTTCGCCGTCGCCGTC[C/G]CTGTCAACCTCGGAGATGAGTTTCC
- Phenotype
-
MIM: 605183
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CALML5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11546426] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CALML5
- Gene Name
- calmodulin like 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017422.4 |
306 |
Missense Mutation |
AGC,AGG |
S58R |
NP_059118.2 |
- Gene
- LOC105376379
- Gene Name
- uncharacterized LOC105376379
There are no transcripts associated with this gene.
View Full Product Details