Product Details

SNP ID
rs139591041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102065573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGGCCGGGGACTCAGCGGTCCG[A/G]GTCCGTGGCAGTCCGGACGGCCGCC
Phenotype
MIM: 607522
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf76 PubMed Links

Gene Details

Gene
C10orf76
Gene Name
chromosome 10 open reading frame 76
There are no transcripts associated with this gene.

Gene
HPS6
Gene Name
HPS6, biogenesis of lysosomal organelles complex 2 subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024747.5 207 Silent Mutation CGA,CGG R33R NP_079023.2

View Full Product Details