Product Details

SNP ID
rs139714298
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:124765431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAGCAACTCTTCCTTGGTCCAAT[C/T]ACATGACGTTATTAGAAAAAAGCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
METTL10 PubMed Links

Gene Details

Gene
METTL10
Gene Name
methyltransferase like 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304467.1 689 Missense Mutation AAT,GAT N115D NP_001291396.1
NM_001304468.1 689 Missense Mutation AAT,GAT N115D NP_001291397.1
NM_212554.3 689 Missense Mutation AAT,GAT N193D NP_997719.2

View Full Product Details