Product Details

SNP ID
rs139813334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:62376760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAACTCACCAAAGAGTTGGCCCA[G/T]AAAACTGCGGAACTGTTGGAAGTTC
Phenotype
MIM: 607818
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC283045 PubMed Links

Gene Details

Gene
LOC283045
Gene Name
uncharacterized LOC283045
There are no transcripts associated with this gene.

Gene
ZNF365
Gene Name
zinc finger protein 365
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014951.2 1430 Missense Mutation CAG,CAT Q189H NP_055766.2
NM_199450.2 1430 Missense Mutation CAG,CAT Q189H NP_955522.1
NM_199451.2 1430 Missense Mutation CAG,CAT Q189H NP_955523.1
NM_199452.3 1430 Intron NP_955524.3
XM_017015937.1 1430 Missense Mutation CAG,CAT Q189H XP_016871426.1
XM_017015938.1 1430 Intron XP_016871427.1
XM_017015939.1 1430 Intron XP_016871428.1
XM_017015940.1 1430 Intron XP_016871429.1
XM_017015941.1 1430 Intron XP_016871430.1

View Full Product Details