Product Details

SNP ID
rs139845028
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:96982082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTGGCATCAGAGGCAGTTTTCA[A/C]TCCTAAGCAGACCCTTACAATTCCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C10orf12 PubMed Links

Gene Details

Gene
C10orf12
Gene Name
chromosome 10 open reading frame 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015652.2 799 Missense Mutation AAT,ACT N231T NP_056467.2

View Full Product Details