Product Details

SNP ID
rs139887829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119168522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGTACTCTTTGGAAGCAGCTGG[A/C]CTTGGCTTGATCTGAAAATACAAAA
Phenotype
MIM: 604769 MIM: 615564
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRDX3 PubMed Links

Gene Details

Gene
PRDX3
Gene Name
peroxiredoxin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302272.1 638 Missense Mutation GGC,GTC G188V NP_001289201.1
NM_006793.4 638 Missense Mutation AGG,AGT R243S NP_006784.1
Gene
SFXN4
Gene Name
sideroflexin 4
There are no transcripts associated with this gene.

View Full Product Details