Product Details

SNP ID
rs139976873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:72097408 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGTGAACCTCTGAGAGATGTGAA[C/T]TGAATTCAGCTTTAGGGAGCCAAAG
Phenotype
MIM: 614215 MIM: 607988
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASCC1 PubMed Links

Gene Details

Gene
ASCC1
Gene Name
activating signal cointegrator 1 complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198798.2 827 Missense Mutation ATT,GTT I334V NP_001185727.1
NM_001198799.2 827 UTR 3 NP_001185728.1
NM_001198800.2 827 Missense Mutation ATT,GTT I334V NP_001185729.1
XM_006717873.3 827 Missense Mutation ATT,GTT I334V XP_006717936.1
XM_011539841.2 827 UTR 3 XP_011538143.1
XM_011539842.2 827 UTR 3 XP_011538144.1
XM_017016291.1 827 UTR 3 XP_016871780.1
XM_017016292.1 827 UTR 3 XP_016871781.1
XM_017016293.1 827 UTR 3 XP_016871782.1
XM_017016294.1 827 UTR 3 XP_016871783.1
XM_017016295.1 827 UTR 3 XP_016871784.1
XM_017016296.1 827 UTR 3 XP_016871785.1
XM_017016297.1 827 UTR 3 XP_016871786.1
XM_017016298.1 827 UTR 3 XP_016871787.1
XM_017016299.1 827 Missense Mutation ATT,GTT I294V XP_016871788.1
XM_017016300.1 827 Missense Mutation ATT,GTT I294V XP_016871789.1
XM_017016301.1 827 UTR 3 XP_016871790.1
XM_017016302.1 827 UTR 3 XP_016871791.1
XM_017016303.1 827 UTR 3 XP_016871792.1
Gene
SPOCK2
Gene Name
sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 2
There are no transcripts associated with this gene.

View Full Product Details