Product Details

SNP ID
rs140551705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103378476 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGACATCTTTGATGGGATGCCG[C/T]GTAGTAAGCAACAGATAGATGCGAT
Phenotype
MIM: 601787
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAF5 PubMed Links

Gene Details

Gene
TAF5
Gene Name
TATA-box binding protein associated factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006951.4 1076 Missense Mutation CGT,TGT R347C NP_008882.2
NM_139052.2 1076 Missense Mutation CGT,TGT R347C NP_620640.1

View Full Product Details