Product Details

SNP ID
rs140627175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:49689149 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGAACCACAGGAGGCAAGACTGC[A/G]GAGGAAGCAAGGAAAGACAGGACTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf53 PubMed Links

Gene Details

Gene
C10orf53
Gene Name
chromosome 10 open reading frame 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042427.1 Intron NP_001035892.1
NM_182554.2 Intron NP_872360.2

View Full Product Details