Product Details
- SNP ID
-
rs140954030
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:69180309 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCCTCGATGTCCTTCTCCCGTGC[C/G]CTATTGTGGGCTCGGCTCCCGGCGG
- Phenotype
-
MIM: 605122
MIM: 605506
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SUPV3L1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12251842] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SUPV3L1
- Gene Name
- Suv3 like RNA helicase
- Gene
- VPS26A
- Gene Name
- VPS26, retromer complex component A
There are no transcripts associated with this gene.
View Full Product Details