Product Details

SNP ID
rs141089126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133160499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATGGACCCGGCCGCGGCGGATC[C/T]TTACGAGGAGGACGGCAAAGACCTG
Phenotype
MIM: 616237
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KNDC1 PubMed Links

Gene Details

Gene
KNDC1
Gene Name
kinase non-catalytic C-lobe domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152643.6 404 Missense Mutation CCT,CTT P11L NP_689856.6
XM_017016858.1 404 Missense Mutation CCT,CTT P11L XP_016872347.1
XM_017016859.1 404 Missense Mutation CCT,CTT P11L XP_016872348.1

View Full Product Details